Order Entry
Netherlands
ContactUsLinkComponent
 
5-Hydroxy-2-pentanone monomer + dimer ≥96.0% (by titrimetric analysis)
 :  TCI
 
undefined
5-Hydroxy-2-pentanone monomer + dimer ≥96.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (Oxime Formation):
    min. 96.0 %

 

Frequently Bought Together