Order Entry
Netherlands
ContactUsLinkComponent
 
Hydroxypropyl cellulose, 1,000-4,000 mPa.s (2% in H₂O 20 °C)
 :  TCI
 
Hydroxypropyl cellulose, 1,000-4,000 mPa.s (2% in H₂O 20 °C)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Solubility in Water:
    almost transparency
  • Drying loss:
    max. 5.0 %
  • Viscosity:
    1,000 to 4,000 mPa-s (2wt%, H2O, 20 deg-C)

 

Frequently Bought Together