Order Entry
Netherlands
ContactUsLinkComponent
Hydroxypropyl cellulose, 6-10 mPa.s (2% in H₂O 20 °C)
Hydroxypropyl cellulose, 6-10 mPa.s (2% in H₂O 20 °C)
Supplier:  TCI
Certificates
Hydroxypropyl cellulose, 6-10 mPa.s (2% in H₂O 20 °C)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Solubility in Water:
    almost transparency
  • Drying loss:
    max. 5.0 %
  • Viscosity:
    6.0 to 10.0 mPa-s (2 wt%, H2O, 20 deg-C)

Specification Test Results

Frequently Bought Together