Order Entry
Netherlands
ContactUsLinkComponent
Hexaethylene glycol monododecyl ether ≥97.0% (by GC)
Hexaethylene glycol monododecyl ether ≥97.0% (by GC)
  TCIAH0444-1ML
 :  TCI
 :  
undefined
Hexaethylene glycol monododecyl ether ≥97.0% (by GC)
  TCIAH0444-1ML
 :  TCI
 :  

 

  • Pk:
    1 mL
  • Seq:
    2070

 

 

  • Appearance:
    White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
  • Purity (GC):
    min. 97.0 %

 

 

 :
 :450.66 g/mol
 :221-282-3
 :3055-96-7
 :C₂₄H₅₀O₇
 :MFCD00042660