Order Entry
Netherlands
ContactUsLinkComponent
 
Heptyl ether ≥98.0% (by GC)
Catalog # TCIAH0364-25ML
Supplier:  TCI
CAS Number:  
Heptyl ether ≥98.0% (by GC)
Catalog # TCIAH0364-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :214.39 g/mol
Boiling Pt:262°C
EINECS:211-100-0
CAS Number:629-64-1
Formula:C₁₄H₃₀O
MDL Number:MFCD00015288