Sodium-4-hydroxyazobenzene-4'-sulphonate hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2623-36-1
:
Formula:C₁₂H₁₀N₂O₄S
MDL Number:MFCD00068861
:300.26 g/mol
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Neutralization titration):min. 98.0 % (calcd.on anh.substance)
- Solubility in hot Water:almost transparency
- Water:max. 13.0 %
Specification Test Results
Frequently Bought Together