Bestelling invoeren
Netherlands
ContactUsLinkComponent
3-Hydroxypropionic acid ∼30% ∼3.6 mol/L in water (contains varying amounts of 3,3'-Oxydipropionic Acid)
Irritant
3-Hydroxypropionic acid ∼30% ∼3.6 mol/L in water (contains varying amounts of 3,3'-Oxydipropionic Acid)
Supplier:  TCI
Certificates
undefined
Irritant
3-Hydroxypropionic acid ∼30% ∼3.6 mol/L in water (contains varying amounts of 3,3'-Oxydipropionic Acid)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Neutralization titration):
    28.0 to 32.0 w/w%

Specification Test Results

Frequently Bought Together