
3-Hydroxy-2-naphthoic acid hydrazide ≥98.0% (by titrimetric analysis)
Catalog # TCIAH0282-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Sodium hydroxide solution:almost transparency
Specification Test Results
About this item
:
:202.21 g/mol
EINECS:226-282-7
:GHS07
CAS Number:5341-58-2
Formula:C₁₁H₁₀N₂O₂
MDL Number:MFCD00004097