
Potassium hydroquinone monosulphate ≥98.0% (by titrimetric analysis)
Catalog # TCIAH0187-10G
Supplier: TCI
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Water:very faint turbidity
Specification Test Results
About this item
:
:228.26 g/mol
EINECS:253-332-5
:GHS07
CAS Number:37067-27-9
Formula:C₆H₆O₅S
MDL Number:MFCD00059841