
Hexadecylsuccinic anhydride ≥95.0% (by GC, titration analysis)
TCIAH0079-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Morpholine Method):min. 95.0 %
- Melting point:74.0 to 78.0 deg-C
- Solubility in hot Toluene:almost transparency
:
:324.51 g/mol
:224-101-6
:GHS07
:4200-91-3
:C₂₀H₃₆O₃
:MFCD00014550