Graphene oxide 10mg/mL dispersion in water 10-30 µm
: TCI
:
:CxOyHz
- Appearance:Red to Dark blue to Black clear liquid to cloudy liquid
- Concentration (with Evaporation residue):9.5 to 10.4 mg/mL
- pH:min. 2 (25 deg-C)
- Average size:10 to 30 micro_m
Frequently Bought Together