Ginsenoside Re ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:52286-59-6
:
Formula:C₄₈H₈₂O₁₈
MDL Number:MFCD00133369
:947.17 g/mol
Storage Temperature:Refrigerator
EINECS:257-814-6
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC) (CAD):min. 98.0 area%
- Elemental analysis (Carbon):57.50 to 63.50 %
Specification Test Results
Frequently Bought Together