
Glimepiride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:93479-97-1
:
Formula:C₂₄H₃₄N₄O₅S
MDL Number:MFCD00878417
:490.62 g/mol
Merck Index:14,04440
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:202.0 to 206.0 deg-C
Specification Test Results
Frequently Bought Together