
Gold's reagent ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:97.0 to 101.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together