Gatifloxacin hydrate ≥98.0% (by HPLC)
Catalog # TCIAG0325-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 95.0 % (calcd.on anh.substance)
- Melting point:180.0 to 184.0 deg-C
- Water:4.5 to 9.0 %
Specification Test Results
About this item
:
:375.40 g/mol
Storage Temperature:Refrigerator
CAS Number:614751-80-3
Formula:C₁₉H₂₂FN₃O₄·xH₂O