Galactinol hydrate ≥99.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:3687-64-7
:
Formula:C₁₂H₂₂O₁₁
:342.30 g/mol
EINECS:222-982-1
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC) (CAD):min. 99.0 area%
- Specific rotation [a]20/D:+128 to +135 deg (C=1, H2O)
- Water (Vaporization methoed):max. 15.0 %
Specification Test Results
Frequently Bought Together