Sodium glycocholate hydrate ≥97.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:338950-81-5
Formula:C₂₆H₄₂NO₆Na·xH₂O
MDL Number:MFCD09037360
:487.60 g/mol (anhy)
Merck Index:14,04494
Storage Temperature:Refrigerator
EINECS:212-730-9
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (with Total Nitrogen):min. 97.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:+26.0 to +30.0 deg (C=1, H2O)
- Water:2.5 to 8.0 %
Specification Test Results
Frequently Bought Together