β-D-Glucose ≥85.0% (by GC) (contains alpha-D-Glucose)
Supplier: TCI
Certificates
About this item
CAS Number:492-61-5
:
Formula:C₆H₁₂O₆
MDL Number:MFCD00063989
Merck Index:14,04459
Storage Temperature:Refrigerator
EINECS:207-756-2
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 85.0 %
- Specific rotation [a]20/D:+50.0 to +56.0 deg (C=10, H2O)
Specification Test Results
Frequently Bought Together