Fmoc-Val-Ala-PAB-OH ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAF1278-250MG
Supplier: TCI
CAS Number:
Specifications
- Pk:250 mg
- Seq:2590
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 96.0 %
- Specific rotation [a]20/D:-26.0 to -30.0 deg (C=1, DMF)
Specification Test Results
About this item
:
:515.61 g/mol
Storage Temperature:Freezer
CAS Number:1394238-91-5
Formula:C₃₀H₃₃N₃O₅
MDL Number:MFCD28557282