Fmoc-Chg-OH ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:161321-36-4
:
Formule:C₂₃H₂₅NO₄
MDL-nummer:MFCD00190878
:379.456 g/mol
Temperatuur bij opslag:Refrigerator
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:181.0 to 185.0 deg-C
- Specific rotation [a]20/D:-3.0 to -140.0 deg (C=1, DMF)
Specificatie testresultaten
Frequently Bought Together