Order Entry
Netherlands
ContactUsLinkComponent
Fmoc-Val-Cit-PAB-OH ≥98.0% (by HPLC)
Fmoc-Val-Cit-PAB-OH ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
Fmoc-Val-Cit-PAB-OH ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results

Frequently Bought Together