Order Entry
Netherlands
ContactUsLinkComponent
2-(4-Fluorophenyl)ethylamine Hydroiodide ≥98.0% (by HPLC, titration analysis)
Irritant
2-(4-Fluorophenyl)ethylamine Hydroiodide ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Irritant
2-(4-Fluorophenyl)ethylamine Hydroiodide ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Argentmetric Titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together