Fmoc-N-Me-Phe-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:77128-73-5
:
Formula:C₂₅H₂₃NO₄
MDL Number:MFCD00151938
:401.462 g/mol
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:132.0 to 136.0 deg-C
- Specific rotation [a]20/D:-58.0 to -62.0 deg (C=1,DMF)
Specification Test Results
Frequently Bought Together