Fmoc-N-Me-Val-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:84000-11-3
:
Formula:C₂₁H₂₃NO₄
MDL Number:MFCD00153395
:353.418 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:188.0 to 192.0 deg-C
- Specific rotation [a]20/D:-68.0 to -72.0 deg (C=1, DMF)
Specification Test Results
Frequently Bought Together