Fmoc-Sar-OH ≥98.0% (by HPLC, titration analysis)
: TCI
:77128-70-2
:
:C₁₈H₁₇NO₄
:MFCD00151926
:311.33 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:117.0 to 121.0 deg-C
Frequently Bought Together