
4-Fluoro-2-(trifluoromethyl)benzyl bromide ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:206860-48-2
:
Formula:C₈H₅BrF₄
MDL Number:MFCD00061176
:257.03 g/mol
Flash Pt:85°C
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together