N-Fmoc-N'-trityl-D-histidine ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:135610-90-1
:
Formula:C₄₀H₃₃N₃O₄
MDL Number:MFCD00077061
:619.721 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Neutralization titration):min. 95.0 %
- Specific rotation [a]20/D:-85.0 to -95.0 deg (C=5, CHCl3)
Specification Test Results
Frequently Bought Together