Order Entry
Netherlands
ContactUsLinkComponent
2,4-Dimethoxy-1-fluorobenzene ≥98.0% (by GC)
2,4-Dimethoxy-1-fluorobenzene ≥98.0% (by GC)
Catalog # TCIAF1139-1G
Supplier:  TCI
CAS Number:  
2,4-Dimethoxy-1-fluorobenzene ≥98.0% (by GC)
Catalog # TCIAF1139-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :156.156 g/mol
Boiling Pt:210°C
Flash Pt:105°C
CAS Number:17715-70-7
Formula:C₈H₉FO₂
MDL Number:MFCD00042265