
2-Fluoro-3-(trifluoromethyl)benzonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:146070-35-1
:
Formula:C₈H₃F₄N
MDL Number:MFCD00061152
Flash Pt:86°C
UN:3276
ADR:6.1,III
:189.11 g/mol
:GHS06
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together