2-Fluoro-4-(trans-4-pentylcyclohexyl)-4'-(trans-4-propylcyclohexyl)biphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:106349-49-9
:
Formula:C₃₂H₄₅F
MDL Number:MFCD11053473
:448.71 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Mesomorphic range:105 to 280 deg-C
Specification Test Results
Frequently Bought Together