3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:306936-57-2
:
Formula:C₁₀H₇FN₂O
MDL Number:MFCD01922131
:190.18 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Acid Value:max. 2.0
Specification Test Results
Frequently Bought Together