Fmoc-L-norvaline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:135112-28-6
:
Formula:C₂₀H₂₁NO₄
MDL Number:MFCD00155631
:339.391 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:153.0 to 157.0 deg-C
- Specific rotation [a]20/D:-20.0 to -22.0 deg (C=2, DMF)
Specification Test Results
Frequently Bought Together