Order Entry
Netherlands
ContactUsLinkComponent
(5-Fluoro-2-nitrophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate ≥95.0% (by HPLC)
Irritant
(5-Fluoro-2-nitrophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate ≥95.0% (by HPLC)
Catalog # TCIAF1110-200MG
Supplier:  TCI
CAS Number:  
undefined
Irritant
(5-Fluoro-2-nitrophenyl)(2,4,6-trimethoxyphenyl)iodonium p-Toluenesulphonate ≥95.0% (by HPLC)
Catalog # TCIAF1110-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 95.0 % (total of Ion-Pair)

Specification Test Results

About this item

 :
 :605.37 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1868173-33-4
Formula:C₂₂H₂₁FINO₈S
MDL Number:MFCD30475654