Order Entry
Netherlands
ContactUsLinkComponent
5'-Fluoro-N-phenyl-[1,1':3',1''-terphenyl]-4'-amine ≥98.0% (by HPLC, total nitrogen)
5'-Fluoro-N-phenyl-[1,1':3',1''-terphenyl]-4'-amine ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAF1070-200MG
Supplier:  TCI
CAS Number:  
5'-Fluoro-N-phenyl-[1,1':3',1''-terphenyl]-4'-amine ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAF1070-200MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    200 mg
  • Seq:
    2530

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light red powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    126.0 to 130.0 deg-C

Specification Test Results

About this item

 :
 :339.41 g/mol
CAS Number:1228153-91-0
Formula:C₂₄H₁₈FN
MDL Number:MFCD27952903