Order Entry
Netherlands
ContactUsLinkComponent
3-Fluoro-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Irritant
3-Fluoro-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAF1064-1G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3-Fluoro-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
Catalog # TCIAF1064-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (Neutralization titration):
    98.0 to 110.0 %

Specification Test Results

About this item

 :
 :207.92 g/mol
Melting Pt:168°C
 :GHS07
CAS Number:159020-59-4
Formula:C₇H₅BF₄O₂
MDL Number:MFCD06657735