
1-(2-Fluorophenyl)-2-thiourea ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:656-32-6
:
Formula:(C₆H₄F)NC(S)NH₂
MDL Number:MFCD00041148
:170.21 g/mol
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:144.0 to 148.0 deg-C
Specification Test Results
Frequently Bought Together