2'-Fluoro-4-pentyl-4''-propyl-1,1':4',1''-terphenyl ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:95759-51-6
:
Formula:C₂₆H₂₉F
MDL Number:MFCD11045061
:360.52 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:51.0 to 55.0 deg-C
- Mesomorphic range:50.0 to 143.0 deg-C
Specification Test Results
Frequently Bought Together