Fmoc-Lys(Ac)-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:159766-56-0
:
Formula:C₂₃H₂₆N₂O₅
MDL Number:MFCD00077409
:410.47 g/mol
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:162.0 to 166.0 deg-C
- Specific rotation [a]20/D:-15.0 to -17.0 deg (C=1,DMF)
Specification Test Results
Frequently Bought Together