Fmoc-L-4-methylphe ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:199006-54-7
:
Formula:C₂₅H₂₃NO₄
MDL Number:MFCD00270197
:401.462 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:169.0 to 173.0 deg-C
- Specific rotation [a]20/D:-31.0 to -34.0 deg (C=1,DMF)
Specification Test Results
Frequently Bought Together