Order Entry
Netherlands
ContactUsLinkComponent
5-Fluoro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Irritant
5-Fluoro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Catalog # TCIAF1026-1G
Supplier:  TCI
CAS Number:  
Irritant
5-Fluoro-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Catalog # TCIAF1026-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    55.0 to 59.0 deg-C

Specification Test Results

About this item

 :
 :223.05 g/mol
 :GHS07
CAS Number:719268-92-5
Formula:C₁₁H₁₅BFNO₂
MDL Number:MFCD07780754