

Fingolimod hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:162359-56-0
:
Formula:C₁₉H₃₄ClNO₂
MDL Number:MFCD00939512
:343.936 g/mol
Merck Index:14,04083
:GHS07
:GHS08
Storage Temperature:Freezer
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together