1-(4-Fluorophenyl)-1,3-butanedione ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:29681-98-9
:
Formula:C₁₀H₉FO₂
MDL Number:MFCD00463139
:180.18 g/mol
Boiling Pt:117°C
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 95.0 %
- Melting point:45.0 to 49.0 deg-C
Specification Test Results
Frequently Bought Together