Fmoc-Cys(tBu)-OH ≥98.0% (by HPLC, titration analysis)
: TCI
:67436-13-9
:
:C₂₂H₂₅NO₄S
:MFCD00037130
:399.505 g/mol
:Refrigerator
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:136.0 to 140.0 deg-C
- Specific rotation [a]20/D:-23.0 to -26.0 deg (C=1,DMF)
Frequently Bought Together