2-(4-Fluorophenyl)-4,5-diphenylimidazole ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAF0968-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:254.0 to 258.0 deg-C
Specification Test Results
About this item
:
:314.36 g/mol
CAS Number:2284-96-0
Formula:C₂₁H₁₅FN₂
MDL Number:MFCD00389667