Fmoc-Inp-OH ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:148928-15-8
:
Formula:C₂₁H₂₁NO₄
MDL Number:MFCD00273475
:351.402 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together