4-(1H-1,2,4-Triazol-1-yl)benzaldehyde ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:27996-86-7
:
Formula:C₉H₇N₃O
MDL Number:MFCD02681969
:173.1714 g/mol
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:147.0 to 151.0 deg-C
Specification Test Results
Frequently Bought Together