N-(4-Fluorobenzylidene)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:5676-81-3
:
Formula:C₁₃H₁₀FN
MDL Number:MFCD00017971
:199.23 g/mol
Boiling Pt:176°C
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:42.0 to 46.0 deg-C
Specification Test Results
Frequently Bought Together