
5'-Formyl-2,2'-bithiophene-5-boronic acid (contains varying amounts of Anhydride)
Catalog # TCIAF0548-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (Neutralization titration):85.0 to 108.0 %
Specification Test Results
About this item
:
:238.08 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1369328-62-0
Formula:C₉H₇BO₃S₂