Order Entry
Netherlands
ContactUsLinkComponent
3-Ferrocenoylpropionic acid ≥95.0% (by GC, titration analysis)
Irritant
3-Ferrocenoylpropionic acid ≥95.0% (by GC, titration analysis)
Catalog # TCIAF0320-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
3-Ferrocenoylpropionic acid ≥95.0% (by GC, titration analysis)
Catalog # TCIAF0320-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Orange to Brown to Dark red powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Purity (Chelometric Titration):
    min. 95.0 %

Specification Test Results

About this item

 :
 :286.11 g/mol
 :GHS07
CAS Number:1291-72-1
Formula:C₁₄H₁₄FeO₃
MDL Number:MFCD00027357