(Ferrocenylmethyl)dodecyldimethylammonium bromide ≥97.0% (by titrimetric analysis)
TCIAF0286-5G
: TCI
:
- Pk:5 g
- Seq:3173
- Appearance:Light yellow to Brown powder to crystal
- Purity (Chelometric Titration):min. 97.0 %
- Melting point:136.0 to 140.0 deg-C
:
:492.37 g/mol
:98778-40-6
:C₂₅H₄₂BrFeN
:MFCD00144308